MilliporeSigma
  • Phenazepam abuse in Finland: findings from apprehended drivers, post-mortem cases and police confiscations.
Phenazepam abuse in Finland: findings from apprehended drivers, post-mortem cases and police confiscations.
Forensic science international (2012-03-07)
Pirkko Kriikku, Lars Wilhelm, Janne Rintatalo, Jukka Hurme, Jan Kramer, Ilkka Ojanperä
ABSTRACT
Phenazepam is a long-acting benzodiazepine that, unlike other benzodiazepines, is currently not scheduled as a narcotic in Finland, most other European countries or the USA. It is used as an anxiolytic, sedative-hypnotic and anti-epileptic, mainly in Russia. In Finland, as well as in some other countries, an increase in the unauthorized use of phenazepam has been observed in recent years. In the one year period between July 1, 2010 and June 30, 2011 the prevalence of phenazepam in Finland was assessed among drivers apprehended for driving under the influence of drugs (DUID), in medico-legal autopsy cases and in police confiscations of illicit drugs. In DUID cases an LC-MS/MS method preceded by solid phase extraction was used for the determination of phenazepam. In the post-mortem investigations the sample preparation consisted of liquid-liquid extraction followed by derivatization and the determination was carried out by GC-MS. The police confiscations were analysed by GC-MS. There were 141 positive phenazepam cases among apprehended drivers, representing approximately 3.5% of all confirmed drug cases (n=4007) in this time period. The median (range) phenazepam blood concentration in DUID cases was 0.061 mg/L (0.004-3.600 mg/L). The median phenazepam concentration in cases with no concomitant stimulant use was significantly higher than the overall median concentration. Phenazepam was found in 17 medico-legal autopsy cases and the median (range) blood concentration was 0.048 mg/L (0.007-1.600 mg/L). Phenazepam was not considered by the medico-legal team to be the sole cause of death in any of the cases, the majority of them being accidental opiod overdoses. There were 26 seizures of phenazepam by the Police in the time period studied, some of the batches consisted of a mixture of phenazepam and stimulant designer drugs. The data show that phenazepam abuse is a widespread phenomenon in Finland. A typical user was a male multi-drug user in his 30s. The concentration range of phenazepam among apprehended drivers and medico-legal autopsy cases was wide and the drug was usually found along with other psychoactive drugs. Therefore, although it seems likely that phenazepam contributed to impairment of driving in some DUID cases, the extent of its effect remains unclear and further studies are needed to define the concentrations causing impairment and toxicity.
Product Description
Supelco
Phenazepam solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®