MilliporeSigma
  • Synthesis of tetrahydro-β-carbolines via isomerization of N-allyltryptamines: a metal-catalyzed variation on the Pictet-Spengler theme.
Synthesis of tetrahydro-β-carbolines via isomerization of N-allyltryptamines: a metal-catalyzed variation on the Pictet-Spengler theme.
Chemical communications (Cambridge, England) (2012-03-01)
Erhad Ascic, Casper L Hansen, Sebastian T Le Quement, Thomas E Nielsen
ABSTRACT
An efficient and broadly applicable alternative to the classical Pictet-Spengler synthesis of tetrahydro-β-carbolines is presented. The method relies on metal-catalyzed isomerization of allylic amines to form reactive iminium intermediates which can be trapped by a tethered indole nucleophile.
Sigma-Aldrich
1,2,3,4-Tetrahydro-9H-pyrido[3,4-b]indole, 98%