MilliporeSigma
  • Short-term testosterone manipulations modulate visual recognition memory and some aspects of emotional reactivity in male rhesus monkeys.
Short-term testosterone manipulations modulate visual recognition memory and some aspects of emotional reactivity in male rhesus monkeys.
Physiology & behavior (2012-03-01)
Agnès Lacreuse, Heather E Gore, Jeemin Chang, Emily R Kaplan
ABSTRACT
The role of testosterone (T) in modulating cognitive function and emotion in men remains unclear. The paucity of animal studies has likely contributed to the slow progress in this area. In particular, studies in nonhuman primates have been lacking. Our laboratory has begun to address this issue by pharmacologically manipulating T levels in intact male rhesus monkeys, using blind, placebo-controlled, crossover designs. We previously found that T-suppressed monkeys receiving supraphysiological T for 4 weeks had lower visual recognition memory for long delays and enhanced attention to videos of negative social stimuli (Lacreuse et al., 2009, 2010) compared to when treated with oil. To further delineate the conditions under which T affects cognition and emotion, the present study focused on the short-term effects of physiological T. Six intact males were treated with the gonadotropin-releasing hormone antagonist degarelix (3 mg/kg) for 7 days and received one injection of T enanthate (5 mg/kg) followed by one injection of oil vehicle 7 days later (n=3), or the reverse treatment (n=3). Performance on two computerized tasks, the Delayed-non-matching-to-sample (DNMS) with random delays and the object-Delayed Recognition Span test (object-DRST) and one task of emotional reactivity, an approach/avoidance task of negative, familiar and novel objects, was examined at baseline and 3-5 days after treatment. DNMS performance was significantly better when monkeys were treated with T compared to oil, independently of the delay duration or the nature (emotional or neutral) of the stimuli. Performance on the object-DRST was unaffected. Interestingly, subtle changes in emotional reactivity were also observed: T administration was associated with fewer object contacts, especially on negative objects, without overt changes in anxious behaviors. These results may reflect increased vigilance and alertness with high T. Altogether, the data suggest that changes in general arousal may underlie the beneficial effects of T on DNMS performance. This hypothesis will require further study with objective measures of physiological arousal.