MilliporeSigma
  • A case of a potential drug interaction between clobazam and etravirine-based antiretroviral therapy.
A case of a potential drug interaction between clobazam and etravirine-based antiretroviral therapy.
Antiviral therapy (2012-02-02)
Mark Naccarato, Deborah Yoong, Colin Kovacs, Kevin Gough
ABSTRACT
The cytochrome P450 isoforms primarily involved in clobazam metabolism are CYP3A4 and 2C19. Drugs that modulate these enzymes would then be expected to alter the exposure of clobazam and its major metabolites. Etravirine, a second-generation non-nucleoside reverse transcriptase inhibitor has been shown to induce CYP3A4, while inhibiting CYP2C9 and CYP2C19. We report a case in which a potential drug interaction between clobazam and etravirine may have led to increased concentrations of clobazam and its pharmacologically active metabolite, N-desmethylclobazam, causing neurotoxic symptoms.
Product Description
Supelco
Clobazam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®