MilliporeSigma
  • Biomedical applications and chemical nature of three dyes first synthesized by Raphael Meldola: isamine blue, Meldola's blue and naphthol green B.
Biomedical applications and chemical nature of three dyes first synthesized by Raphael Meldola: isamine blue, Meldola's blue and naphthol green B.
Biotechnic & histochemistry : official publication of the Biological Stain Commission (2011-12-14)
M Hope-Roberts, R W Horobin
ABSTRACT
Brief accounts are given of the chemical nature, and past and current biomedical applications of three dyes first synthesized by Raphael Meldola: isamine blue, Meldola's blue and naphthol green B.