MilliporeSigma
  • In situ trace analysis of oil in water with mid-infrared fiberoptic chemical sensors.
In situ trace analysis of oil in water with mid-infrared fiberoptic chemical sensors.
Analytical chemistry (2011-11-23)
Yuliya Luzinova, Bogdan Zdyrko, Igor Luzinov, Boris Mizaikoff
ABSTRACT
The determination of trace amounts of oil in water facilitates the forensic analysis on the presence and origin of oil in the aqueous environment. To this end, the present study focuses on direct sensing schemes for quantifying trace amounts of oil in water using mid-infrared (MIR) evanescent field absorption spectroscopy via fiberoptic chemical sensors. MIR transparent silver halide fibers were utilized as optical transducer for interrogating oil-in-water emulsions via the evanescent field emanating from the waveguide surface, and penetrating the surrounding aqueous environment by a couple of micrometers. Unmodified fibers and fibers surface-modified with grafted epoxidized polybutadiene layers enabled the direct detection of crude oil in a deionized water matrix at the ppm level to ppb concentration level, respectively. Thus, direct chemical sensing of crude oil IR signatures without any sample preparation as low as 46 ppb was achieved with a response time of a few seconds.
Sigma-Aldrich
Silver bromide, 99%