MilliporeSigma
  • [Proteolysis of semax analogues with different N-terminal amino acids by aminopeptidases].
[Proteolysis of semax analogues with different N-terminal amino acids by aminopeptidases].
Bioorganicheskaia khimiia (2011-11-22)
K V Shevchenko, T V V'iunova, I Iu Nagaev, L A Andreeva, L Iu Alfeeva, N F Miasoedov
ABSTRACT
Proteolysis of semax (Met-Glu-His-Phe-Pro-Gly-Pro, Sem) and its analogues ([Ala1]Sem, [Gly1]Sem, [Thr1]Sem, [Trp1]Sem) that are differ from semax in substitution of N-terminal Met residue were studied. It is shown that such replacement changes the rate of peptides degradation by N-aminopeptidases (EC 3.4.11.2, Sigma, Type VI, 9.2 units. Akt. / mg). [Ala1]Sem, [Gly1]Sem and [Thr1]Sem semax analogues proved to be more stable to proteolysis than semax (Sem), and their initial product of proteolysis is His-Phe-Pro-Gly-Pro (Sem-5). For triptophan analogue both Glu-His-Phe-Pro-Gly-Pro (Sem-6) and Sem-5 product are formed in similar quantities. It is found that all investigated analogues can be used as inhibitors in Sem proteolysis.
Sigma-Aldrich
Semax, ≥98% (HPLC)