MilliporeSigma
  • N-acetylaspartylglutamate (NAAG) and N-acetylaspartate (NAA) in patients with schizophrenia.
N-acetylaspartylglutamate (NAAG) and N-acetylaspartate (NAA) in patients with schizophrenia.
Schizophrenia bulletin (2011-09-15)
Frank Jessen, Natascha Fingerhut, Alois M Sprinkart, Kai-Uwe Kühn, Nadine Petrovsky, Wolfgang Maier, Hans-H Schild, Wolfgang Block, Michael Wagner, Frank Träber
ABSTRACT
BACKGROUND : Imbalance of glutamatergic neurotransmission has been proposed as a key mechanism underlying symptoms of schizophrenia. The neuropetide N-acetylaspartylglutamate (NAAG) modulates glutamate release. NAAG provides a component of the proton magnetic resonance spectrum (1H-MRS) in humans. The signal of NAAG, however, largely overlaps with its precursor and degrading product N-acetylaspartate (NAA) that by itself does not act in glutamatergic neurotransmission. We quantified NAAG and NAA separately from the 1H-MRS signal in 20 patients with schizophrenia and 20 healthy comparison subjects on a 3.0 Tesla MR scanner. The 1H-MRS voxels were positioned in the anterior cingulate cortex (ACC) and in the left frontal lobe. Psychopathological symptoms and cognitive performance were assessed. In the ACC, the ratio NAAG/NAA was increased (P = .041) and NAAG was increased at a trend level (P = .066) in patients, while NAA was reduced (P = .030). NAA correlated with attention performance in patients (r = .64, P = .005) in the ACC. There was no group difference of NAAG, NAA, or NAAG/NAA in the frontal lobe but an inverse correlation of NAAG with negatives symptoms (Positive and Negative Symptoms Scale [PANSS] negative, r = -.58, P = .018) and with the total symptom score (PANSS total, r = -.50, P = .049). In addition, there was a positive correlation of frontal lobe NAAG (r = .53, P = .035) and NAAG/NAA (r = .54, P = .030) with episodic memory in patients. In this study, we present the first in vivo evidence for altered NAAG concentration in patients with schizophrenia.
Sigma-Aldrich
N-Acetyl-Asp-Glu, ≥97% (TLC)