Merck
  • Pollution of soils with organophosphorus flame retardants and plasticizers.
Pollution of soils with organophosphorus flame retardants and plasticizers.
Journal of environmental monitoring : JEM (2011-08-27)
Elke Fries, Ivana Mihajlović
ABSTRACT
The detection of the three organophosphate esters TCEP, TCPP and TBEP in soil samples indicates that pollution of soils from diffuse atmospheric sources has to be considered in risk assessments.
Sigma-Aldrich
4,4′,4′′,4′′′-(Porphine-5,10,15,20-tetrayl)tetrakis(benzoic acid), Dye content 75 %
Pricing and availability is not currently available.