MilliporeSigma
  • Net oxidative addition of C(sp3)-F bonds to iridium via initial C-H bond activation.
Net oxidative addition of C(sp3)-F bonds to iridium via initial C-H bond activation.
Science (New York, N.Y.) (2011-06-28)
Jongwook Choi, David Y Wang, Sabuj Kundu, Yuriy Choliy, Thomas J Emge, Karsten Krogh-Jespersen, Alan S Goldman
ABSTRACT
Carbon-fluorine bonds are the strongest known single bonds to carbon and as a consequence can prove very hard to cleave. Alhough vinyl and aryl C-F bonds can undergo oxidative addition to transition metal complexes, this reaction has appeared inoperable with aliphatic substrates. We report the addition of C(sp(3))-F bonds (including alkyl-F) to an iridium center via the initial, reversible cleavage of a C-H bond. These results suggest a distinct strategy for the development of catalysts and promoters to make and break C-F bonds, which are of strong interest in the context of both pharmaceutical and environmental chemistry.