MilliporeSigma
  • Enhanced anti-tumor effect of 9-nitro-camptothecin complexed by hydroxypropyl-β-cyclodextrin and safety evaluation.
Enhanced anti-tumor effect of 9-nitro-camptothecin complexed by hydroxypropyl-β-cyclodextrin and safety evaluation.
International journal of pharmaceutics (2011-06-08)
Ye Jiang, Xinyi Jiang, Kitki Law, Yanzuo Chen, Jijin Gu, Wei Zhang, Hongliang Xin, Xianyi Sha, Xiaoling Fang
ABSTRACT
The aim of this study was to evaluate the safety and anti-tumor effect of 9-nitro-camptothecin/hydroxypropyl-β-cyclodextrin (9-NC/HP-β-CD) complex on tumor-bearing mice. The in vitro anti-tumor activity was tested by MTT assay. Our study revealed that the 9-NC/HP-β-CD complex showed significant anti-tumor activity towards Skov-3, MCF-7, HeLa and S180 cell lines with IC(50) values of 0.24 ± 0.09, 0.59 ± 0.20, 0.83 ± 0.11, and 6.30 ± 2.42 μg/ml, respectively, significantly superior to the free 9-NC. The in vivo therapeutic efficacy was investigated in ICR mice bearing mouse sarcoma S180. Both the high (3mg/kg) and low (1mg/kg) doses of 9-NC/HP-β-CD complex demonstrated high inhibition ratio of tumor growth (>75%). The subacute toxicity test was performed by measuring the body weight, histopathology, blood cell counts and clinical chemistry parameters (total bilirubin, alanine transferase, aspartate transferase, blood urea nitrogen and creatinine), and the results indicated the good safety profile of the complex. Taken together, the results suggested that the 9-NC complexed in HP-β-CD, instead of dissolved in the organic solvent, presented significant anti-tumor activity and low toxicity for the treatment of cancer.
Sigma-Aldrich
Rubitecan