MilliporeSigma
  • Deciphering the complexity of sainfoin (Onobrychis viciifolia) proanthocyanidins by MALDI-TOF mass spectrometry with a judicious choice of isotope patterns and matrixes.
Deciphering the complexity of sainfoin (Onobrychis viciifolia) proanthocyanidins by MALDI-TOF mass spectrometry with a judicious choice of isotope patterns and matrixes.
Analytical chemistry (2011-04-15)
Elisabetta Stringano, Rainer Cramer, Wayne Hayes, Celia Smith, Trevor Gibson, Irene Mueller-Harvey
ABSTRACT
Use of superdihydroxybenzoic acid as the matrix enabled the analysis of highly complex mixtures of proanthocyanidins from sainfoin (Onobrychis viciifolia) by MALDI-TOF mass spectrometry. Proanthocyanidins contained predominantly B-type homopolymers and heteropolymers up to 12-mers (3400 Da). Use of another matrix, 2,6-dihydroxyacetophenone, revealed the presence of A-type glycosylated dimers. In addition, we report here how a comparison of the isotopic adduct patterns, which resulted from Li and Na salts as MALDI matrix additives, could be used to confirm the presence of A-type linkages in complex proanthocyanidin mixtures. Preliminary evidence suggested the presence of A-type dimers in glycosylated prodelphinidins and in tetrameric procyanidins and prodelphinidins.
Product Description
Supelco
2′,6′-Dihydroxyacetophenone, matrix substance for MALDI-MS, ≥99.5%
Supelco
2′,6′-Dihydroxyacetophenone, matrix substance for MALDI-MS, ≥99.5% (HPLC), Ultra pure
Sigma-Aldrich
2′,6′-Dihydroxyacetophenone, 97%