MilliporeSigma
  • All-trans retinoic acid and a novel synthetic retinoid tamibarotene (Am80) differentially regulate CD38 expression in human leukemia HL-60 cells: possible involvement of protein kinase C-delta.
All-trans retinoic acid and a novel synthetic retinoid tamibarotene (Am80) differentially regulate CD38 expression in human leukemia HL-60 cells: possible involvement of protein kinase C-delta.
Journal of leukocyte biology (2011-03-12)
Akira Uruno, Naoya Noguchi, Ken Matsuda, Koji Nata, Takeo Yoshikawa, Youichiro Chikamatsu, Hiroyuki Kagechika, Hideo Harigae, Sadayoshi Ito, Hiroshi Okamoto, Akira Sugawara
ABSTRACT
ATRA and a synthetic RAR agonist tamibarotene (Am80) induce granulocytic differentiation of human acute leukemia HL-60 cells and have been used in antineoplastic therapy. ATRA induces CD38 antigen during HL-60 cell differentiation, which interacts with CD31 antigen on the vascular EC surface and may induce disadvantages in the therapy. We here examined the mechanisms of the ATRA-mediated CD38 induction and compared the difference between ATRA- and tamibarotene-mediated induction. Tamibarotene-induced HL-60 cell adhesion to ECs was 38% lower than ATRA, and NB4 cell adhesion to ECs by tamibarotene was equivalent to ATRA, which induced CD38 gene transcription biphasically in HL-60 cells, the early-phase induction via DR-RARE containing intron 1, and the delayed-phase induction via RARE lacking the 5'-flanking region. In contrast to ATRA, tamibarotene induced only the early-phase induction, resulting in its lower CD38 induction than ATRA. A PKCδ inhibitor, rottlerin, and siRNA-mediated PKCδ knockdown suppressed the ATRA-induced CD38 promoter activity of the 5'-flanking region, whereas a RAR antagonist, LE540, or RAR knockdown did not affect it. Cycloheximide and rottlerin suppressed the delayed-phase induction of CD38 expression by ATRA but did not affect the early-phase induction. Moreover, ATRA, but not tamibarotene, induced PKCδ expression without affecting its mRNA stability. The diminished effect of tamibarotene on CD38-mediated HL-60 cell adhesion to ECs compared with ATRA is likely a result of the lack of its delayed-phase induction of CD38 expression, which may be advantageous in antineoplastic therapy.
Sigma-Aldrich
Tamibarotene, ≥98% (HPLC)