MilliporeSigma
  • High frequency vibrational modulations in two-dimensional electronic spectra and their resemblance to electronic coherence signatures.
High frequency vibrational modulations in two-dimensional electronic spectra and their resemblance to electronic coherence signatures.
The journal of physical chemistry. B (2011-02-19)
N Christensson, F Milota, J Hauer, J Sperling, O Bixner, A Nemeth, H F Kauffmann
ABSTRACT
In this work we analyze how nuclear coherences modulate diagonal and off-diagonal peaks in two-dimensional electronic spectroscopy. 2D electronic spectra of pinacyanol chloride are measured with 8 fs pulses, which allows coherent excitation of the 1300 cm(-1) vibrational mode. The 2D spectrum reveals both diagonal and off-diagonal peaks related to the vibrational mode. On early time scales, up to 30 fs, coherent dynamics give rise to oscillations in the amplitudes, positions, and shapes of the peaks in the 2D spectrum. We find an anticorrelation between the amplitude and the diagonal width of the two diagonal peaks. The measured data are reproduced with a model incorporating a high frequency mode coupled to an electronic two-level-system. Our results show that these anticorrelated oscillations occur for vibrational wavepackets and not exclusively for electronic coherences as has been assumed previously.
Sigma-Aldrich
Pinacyanol chloride
Sigma-Aldrich
1,1′-Diethyl-2,2′-carbocyanine iodide, 97%