MilliporeSigma
  • Synthesis of 7-aza-5-deazapurine analogues via copper(I)-catalyzed hydroamination of alkynes and 1-iodoalkynes.
Synthesis of 7-aza-5-deazapurine analogues via copper(I)-catalyzed hydroamination of alkynes and 1-iodoalkynes.
The Journal of organic chemistry (2010-11-26)
Larissa B Krasnova, Jason E Hein, Valery V Fokin
ABSTRACT
A new method for the synthesis of dihydroimidazo[1,2-a][1,3,5]triazin-4(6H)-ones via copper(I)-catalyzed hydroamination was developed. In addition, for the first time, iodoalkynes were shown to participate in the copper(I)-catalyzed intramolecular hydroamination reaction with exclusive formation of E-isomers.
Sigma-Aldrich
4-Azabenzimidazole, 99%