MilliporeSigma
  • [Beta-adrenoblockers for the correction of arterial hypertension in postmenopausal women].
[Beta-adrenoblockers for the correction of arterial hypertension in postmenopausal women].
Klinicheskaia meditsina (2010-11-26)
I A Latfullin, G P Ishmurzin, R F Gaĭfullina, R I Akhmerova, Z F Kim, A A Podol'skaia
ABSTRACT
The use of beta-blockers is an important component of therapy of cardiovascular pathology (e.g. coronary heart disease, arterial hypertension) in menopausal women. Comparative data on the efficiency of lokren and carvedilol for the correction of grade 2 AH are presented.
Sigma-Aldrich
Betaxolol hydrochloride, >98% (HPLC)