MilliporeSigma
  • A novel terpyridine/benzofurazan hybrid fluorophore: metal sensing behavior and application.
A novel terpyridine/benzofurazan hybrid fluorophore: metal sensing behavior and application.
Dalton transactions (Cambridge, England : 2003) (2010-11-23)
Zhenghao Yang, Chongchong Yan, Yuncong Chen, Chengcheng Zhu, Changli Zhang, Xindian Dong, Weiqi Yang, Zijian Guo, Yi Lu, Weijiang He
ABSTRACT
Terpyridine/benzofurazan conjugation results in a new hybrid fluorophore of the colorimetric sensing ability for Fe(2+) and fluorimetric sensing ability for XII group cations. The improved emission properties and cell imaging ability imply it is a suitable platform to construct a fluorescent sensor for metal imaging in biological systems.
Sigma-Aldrich
Benzofurazan, 97%