MilliporeSigma
  • Parameter optimization of the fungicide (Vapam) sorption onto soil modified with clinoptilolite by Taguchi method.
Parameter optimization of the fungicide (Vapam) sorption onto soil modified with clinoptilolite by Taguchi method.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes (2010-10-23)
Seyed N Azizi, Neda Asemi
ABSTRACT
This study employs the Taguchi optimization methodology to optimize the effective parameters for the pesticide (Vapam) sorption onto soil modified with natural zeolite (clinoptilolite). The experimental factors and their ranges chosen for determination of the effective parameters were: initial Vapam concentration (0.4-1.6 mg/L), initial pH of the pesticide solution (2-12), the percentage of clinoptilolite in the modified soil (0-6 %), temperature (15-35°C) and shaking time (2-24 h). The orthogonal array (OA) L(16) and the bigger the better response category of the Taguchi method were selected to determine the optimum conditions: initial Vapam concentration (1.2 mg/L), initial pH of the pesticide solution (2), the percentage of clinoptilolite in the modified soil (4 %), temperature (15°C) and shaking time (2 h). The results showed that in comparison with other parameters, the initial Vapam concentration was the most effective one for the sorption of this pesticide onto soil, modified with clinoptilolite. Moreover, after determining the optimum levels of the sorption process parameters, confirmation experiments were performed to prove the effectiveness of the Taguchi's experimental design methodology.
Product Description
Supelco
Metam-sodium hydrate, PESTANAL®, analytical standard