MilliporeSigma
  • Establishment, characterization, and drug sensitivity of a new Ewing sarcoma cell line (SS-ES-1).
Establishment, characterization, and drug sensitivity of a new Ewing sarcoma cell line (SS-ES-1).
Journal of pediatric hematology/oncology (2010-10-22)
Saeed Rajabalian, Zahra Hajializadeh, Iran Pooraboli, Hamid Jangi-Aghdam, Marziyeh Badinloo
ABSTRACT
Ewing sarcoma (ES) is one of the most malignant tumors in children and young adults. We present here a new ES cell line, SS-ES-1, established from the left thoracic tumor of a 16-year-old female patient. The SS-ES-1 cells retained genotype, morphology, and growth rate for over 150 passages. Immunocytochemical staining showed the strong immunoreactivity for cytokeratin, epithelial membrane antigen, neurofilament, CD99, P53, Ki-67, platelet-derived growth factor receptor-β, estrogen receptor-α (ER-α), and Bcl-2, but no reactivity for glial fibrillary acidic protein, epidermal growth factor receptor, and HER-2/neu. The presence of the type 1 EWS/FLI-1 fusion transcripts was confirmed by reverse transcriptase-polymerase chain reaction. On the basis of the MTT assay results, GW2974, a dual inhibitor of epidermal growth factor receptor and HER-2/neu, exhibited only a weak cytotoxic response in SS-ES-1 cells. In contrast, tyrphostin A9, a specific inhibitor of platelet-derived growth factor receptor, had a high cytotoxic effect against these cells. Surprisingly, it was found that SS-ES-1 cells displayed a high sensitivity to 4OH-tamoxifen. In conclusion, the SS-ES-1 cell line shows unique cellular properties, which makes it a useful model for studying various aspects of the biology of ES. In addition, the results suggest that ER can be a good therapeutic target for ER + ES.
Sigma-Aldrich
Tyrphostin A9, solid