MilliporeSigma
  • Study on the effect of etofenamate 10% cream in comparison with an oral NSAID in strains and sprains due to sports injuries.
Study on the effect of etofenamate 10% cream in comparison with an oral NSAID in strains and sprains due to sports injuries.
Acta Belgica. Medica physica : organe officiel de la Societe royale belge de medecine physique et de rehabilitation (1990-07-01)
G Vanderstraeten, P Schuermans
ABSTRACT
In this 60 patient study of sports traumatology due to football injuries, etofenamate gel proved equally effective as oral naproxen on the overall pain scores (65% none to mild pain for etofenamate versus 86% for naproxen; p greater than 0.05). The global clinical impression results have been rated as good or excellent in 44% in the naproxen group versus 50% in the etofenamate group. The incidence of side effects in the etofenamate group (3%) was lower than in the naproxen group (20%). This study demonstrates that etofenamate gel has equal efficacy as oral NSAIDS but a better side effect profile in sport injuries in football players.
Product Description
Etofenamate, European Pharmacopoeia (EP) Reference Standard