MilliporeSigma
  • Synthesis of continuous TiC nanofibers and/or nanoribbons through electrospinning followed by carbothermal reduction.
Synthesis of continuous TiC nanofibers and/or nanoribbons through electrospinning followed by carbothermal reduction.
Nanoscale (2010-09-08)
Lifeng Zhang, Jianjun Hu, Andrey A Voevodin, Hao Fong
ABSTRACT
Continuous titanium carbide (TiC) nanofibers that possess an intriguing nanoribbon morphology with a width and thickness of approximately 300 nm and approximately 40 nm, respectively, and containing TiC crystallites with sizes ranging from 5 nm to 30 nm were synthesized through electrospinning followed by carbothermal reduction.
Sigma-Aldrich
Titanium(IV) carbide, powder, <4 μm, ≥99% (Ti)
Sigma-Aldrich
Titanium carbide, nanopowder, ≤200 nm particle size (SEM)
Sigma-Aldrich
Polyvinylpyrrolidone, average Mw ~1,300,000 by LS
Sigma-Aldrich
Titanium(IV) carbide, −325 mesh, 98%
Sigma-Aldrich
Furfuryl alcohol, 98%