MilliporeSigma
  • Composition and antimicrobial and anti-wood-decay fungal activities of the leaf essential oils of Machilus pseudolongifolia from Taiwan.
Composition and antimicrobial and anti-wood-decay fungal activities of the leaf essential oils of Machilus pseudolongifolia from Taiwan.
Natural product communications (2010-08-26)
Chen-Lung Ho, Pei-Chun Liao, Kuang-Ping Hsu, Eugene I-Chen Wang, Wei-Chih Dong, Yu-Chang Su
ABSTRACT
The hydrodistillated leaf essential oil of Machilus pseudolongifolia was analyzed to determine its composition and yield. Seventy compounds were identified, the main components being beta-eudesmol (26.8%), alpha-cadinol (20.8%), viridiflorene (8.9%), alpha-caryophyllene (5.3%), globulol (4.6%) and beta-caryophyllene (4.2%). Oxygenated sesquiterpenes (60.1%) and sesquiterpene hydrocarbons (31.4%) were the predominant groups of compounds. The leaf oil exhibited excellent antimicrobial and anti-wood-decay fungal activities.
Sigma-Aldrich
β-Eudesmol, ≥90% (GC)