MilliporeSigma
  • The role of serotonin-2 (5-HT2) and dopamine receptors in the behavioral actions of the 5-HT2A/2C agonist, DOI, and putative 5-HT2C inverse agonist, SR46349B.
The role of serotonin-2 (5-HT2) and dopamine receptors in the behavioral actions of the 5-HT2A/2C agonist, DOI, and putative 5-HT2C inverse agonist, SR46349B.
Psychopharmacology (2010-07-24)
Laura C Scarlota, John A Harvey, Vincent J Aloyo
ABSTRACT
Atypical antipsychotic efficacy is often attributed to actions at serotonin-2 (5-HT(2)) and dopamine receptors, indicating a potential benefit of understanding the interplay between these systems. Currently, it is known that 5-HT(2) receptors modulate dopamine release, although the role of specific dopamine receptors in 5-HT(2)-mediated behavior is not well understood. We examined the role of 5-HT(2A), 5-HT(2C), and dopamine (D1 and D2) receptors in the behavioral response to a 5-HT(2A/2C) agonist (DOI) and 5-HT(2A/2C) antagonist (SR46349B). Effects were assessed by measuring rabbit head bobs (previously characterized as 5-HT(2A) receptor-mediated) and body shakes (5-HT(2C)-mediated). As expected, DOI produced head bobs and body shakes, and these DOI-elicited behaviors were attenuated by the SR46349B pretreatment. Unexpectedly, SR46349B also induced head bobs when administered alone. However, SR46349B-elicited head bobs are distinguishable from those produced by DOI since the 5-HT(2A) antagonist, ketanserin, only attenuated DOI-elicited head bobs. Conversely, 5-HT(2C) ligands (SB242084 and SB206553) inhibited SR46349B but not DOI-induced head bobs. Furthermore, when administered alone, SB206553 (a 5-HT(2C) inverse agonist) produced head bobs, indicating the behavior can be either 5-HT(2A) or 5-HT(2C) mediated. Next, it was revealed that D1 and D2 receptors play a role in DOI-elicited head bobs, but only D1 receptors are required for SR46349B-elicited head bobs. 5-HT(2A) receptor agonism and 5-HT(2C) inverse agonism produce the same behavior, likely due to similar downstream actions at D1 receptors. Consequently, 5-HT(2C) agonism or D1 agonism may be effective therapies for disorders, such as schizophrenia, currently being treated with 5-HT(2A) antagonists.
Sigma-Aldrich
SR-46349 hemifumarate salt, ≥98% (HPLC)