MilliporeSigma
  • Al3+ selective an efficient colorimetric receptor derived from 5-aminouracil.
Al3+ selective an efficient colorimetric receptor derived from 5-aminouracil.
Talanta (2010-07-07)
K K Upadhyay, Ajit Kumar
ABSTRACT
An interference-free naked-eye recognition of Al(3+) at its micromolar level has been done in 5% aqueous DMSO solution employing a Schiff base 5-[(2-hydroxy-5-nitro-benzylidene)-amino]-1H-pyrimidine-2,4-dione (receptor 1) which is an intramolecular charge transfer (ICT) probe. The pyrimidine and nitrophenyl groups serve as electron rich (donor) and deficient (acceptor) pockets in receptor 1 exhibiting a broad ICT band at 434 nm (olive green). The concomitant additions of Al(3+) as its chloride salt to the 5x10(-5) M aqueous DMSO solution of the receptor 1 lead hypsochromic shifting of its ICT band to 395 nm (colorless). The same ICT band undergoes a marginal bathochromic shifting (6 nm) along with a hyperchromic shift on separate additions of a basic anion like F(-), CH(3)COO(-) and H(2)PO(4)(-) to the receptor 1 and faced almost similar fate on concomitant additions of Al(3+) as mentioned above.
Sigma-Aldrich
5-Aminouracil, 98%