MilliporeSigma
  • Gas-phase distribution and emission of chloropicrin applied in gelatin capsules to soil columns.
Gas-phase distribution and emission of chloropicrin applied in gelatin capsules to soil columns.
Journal of environmental quality (2010-04-20)
Qiuxia Wang, Dong Wang, Jueting Tang, Dongdong Yan, Hongjun Zhang, Fangyan Wang, Meixia Guo, Aocheng Cao
ABSTRACT
Chloropicrin (CP) is highly volatile and may pose strong exposure risks for humans and the environment. A gelatin capsule formulation was developed to reduce atmospheric CP emissions and to improve application safety. The objective of this study was to determine the distribution, atmospheric emissions, and soil residual of CP after application in gelatin capsules to soil columns. Two treatments were studied: (i) CP liquid injection with polyethylene film and (ii) CP gelatin capsules with polyethylene film. For the CP liquid injection treatment, the concentration of CP peaked (120.7 microg cm(-3)) at the 20-cm depth 1 h after fumigant injection and decreased with time; at other depths, the CP concentration increased initially and decreased after a typical diffusion-dispersion process. For the gelatin capsule treatment, concentrations of CP at 20 cm depth increased slowly and peaked at 30.04 microg cm(-3) 264 h after application, indicating a slow CP release through the gelatin capsule shells. The results indicate that a rapid breakdown of gelatin capsules occurred between 11 and 12 d after application. The application of CP gelatin capsules reduced total CP emission by approximately 3 times compared with liquid injection with film cover. Similar residual soil CP was found between the liquid injection and the two gelatin capsule treatments. Chloropicrin gelatin capsules could be a promising new technology for reducing environmental emissions and potential human exposure.