MilliporeSigma
  • 15-Lipoxygenases and its metabolites 15(S)-HETE and 13(S)-HODE in the development of non-small cell lung cancer.
15-Lipoxygenases and its metabolites 15(S)-HETE and 13(S)-HODE in the development of non-small cell lung cancer.
Thorax (2010-04-15)
Huiling Yuan, Ming-Yue Li, Lily T Ma, Michael K Y Hsin, Tony S K Mok, Malcolm J Underwood, George G Chen
ABSTRACT
15-S-Hydroxyeicosatetraenoic acid (15(S)-HETE) and 13-S-hydroxyoctadecadienoic acid (13(S)-HODE), both of which are metabolites of 15-lipoxygenases (15-LOXs), are endogenous ligands for peroxisome proliferator-activated receptor gamma (PPARgamma). The activation of PPARgamma inhibits cell growth and induces apoptosis in some cancers. The role of 15(S)-HETE) and 13(S)-HODE in the development of lung cancer is not clear. 15-LOXs, 15(S)-HETE and 13(S)-HODE were monitored during the development of mouse lung tumours induced by the tobacco smoke carcinogen 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanone (NNK) and the levels of these markers were determined in 54 human non-small cell lung cancers. 15-LOXs, 15(S)-HETE and 13(S)-HODE levels were significantly reduced in human lung cancer tissue compared with non-tumour lung tissue (p=0.011 and p=0.022, respectively). In mouse experiments, 15(S)-HETE and 13(S)-HODE started to reduce at 26 and 30 weeks, respectively, after NNK treatment. The time frame of 15(S)-HETE reduction was in line with the decrease in 12/15-LOX mRNA and protein. A significant difference in the number of tumours in NNK-treated mice and controls was not observed until week 34 (p<0.05) and week 38 (p<0.01). The reduction in 12/15-LOX and 15(S)-HETE therefore predated the appearance of lung tumour. Furthermore, PPARgamma activity was decreased in NNK-treated mouse lungs compared with the control, and the decreased PPARgamma activity occurred at the same time points as the reduction in 12/15-LOX and 15(S)-HETE. These findings indicate that the reduction in 15-LOX, 15(S)-HETE and 13(S)-HODE results in the decreased PPARgamma activity seen in lung tumours and contributes to the development of lung tumours induced by tobacco smoking.
Product Description
Avanti
15(S)-HETE, Avanti Research - A Croda Brand, ethanol solution
Avanti
13(S)HODE, Avanti Research - A Croda Brand (900130E), ethanol solution