MilliporeSigma
[A case report of alverine-citrate-induced acute hepatitis].
The Korean journal of hepatology (2010-04-09)
Jee Young Han, Jin Woo Lee, Joon Mee Kim, Kowoon Joo, Ung Chon, Jung Il Lee, Seok Jeong, Don Haeng Lee, Young Soo Kim, Kyung Sun Min
ABSTRACT
Alverine citrate is one of the most commonly used antispasmodic drugs for patients with irritable bowel syndrome. Alverine-citrate-induced hepatotoxicity is extremely rare, with only a few cases having been reported worldwide. We present a case of a 75-year-old female patient who experienced complicated jaundice and abdominal discomfort after taking alverine citrate. Other causes of hepatitis were ruled out and the results of the liver function test returned to normal after ceasing the drug. This is the first case report in Korea of alverine-citrate-induced hepatotoxicity.
Sigma-Aldrich
Alverine citrate salt
Alverine citrate salt, European Pharmacopoeia (EP) Reference Standard