MilliporeSigma
  • Biotransformation of artemisinin using cell suspension cultures of Catharanthus roseus (L.) G.Don and Lavandula officinalis L.
Biotransformation of artemisinin using cell suspension cultures of Catharanthus roseus (L.) G.Don and Lavandula officinalis L.
Biotechnology letters (2010-04-07)
Suman Patel, Rashmi Gaur, Priyanka Verma, Rajendra S Bhakuni, Archana Mathur
ABSTRACT
Artemisinin, an antimalarial compound, at 5 mg/40 ml, was transformed by cell suspension cultures of Catharanthus roseus (L.) G.Don and Lavandula officinalis L. into deoxyartemisinin with yields >78% (3.93 mg deoxyartemisinin from 5 mg artemisinin). Maximum conversion (78.6 and 78%) occurred after 6 and 7 days of adding artemisinin to 20 and 9 days old cultures of C. roseus and L. officinalis, respectively. The procedure was scaled up by and 500 mg artemisinin was transformed into 390 mg deoxyartemisinin. Addition of artemisinin at the beginning of the culture cycle resulted in >50% reduction in dry biomass production with no bioconversion. Conversion of artemisinin occurred intracellularly followed by leaching of the product into the medium.
Sigma-Aldrich
(3R,3aS,6R,6aS,9S,10aS,10bR)-Octahydro-3,6,9-trimethyl-10aH-9,10b-epoxypyrano[4,3,2-jk][2]benzoxepin-2(3H)-one, AldrichCPR