MilliporeSigma
  • Attenuation of Vibrio fischeri quorum sensing using rationally designed polymers.
Attenuation of Vibrio fischeri quorum sensing using rationally designed polymers.
Biomacromolecules (2010-03-17)
Elena V Piletska, Georgios Stavroulakis, Kal Karim, Michael J Whitcombe, Iva Chianella, Anant Sharma, Kevin E Eboigbodin, Gary K Robinson, Sergey A Piletsky
ABSTRACT
A first attempt to attenuate the quorum sensing (QS) of a marine heterotroph microorganism, Vibrio fischeri , using signal molecule-sequestering polymers (SSPs) is presented. A set of rationally designed polymers with affinity toward a signal molecule of V. fischeri , N-(beta-ketocaproyl)-l-homoserine lactone (3-oxo-C6-AHL) was produced. It is reported that computationally designed polymers could sequester a signal molecule of V. fischeri and prevent QS-controlled phenotypes (in this case, bioluminescence) from being up-regulated. It was proven that the attenuation of bioluminescence of V. fischeri was due to sequestration of the signal molecule by specific polymers and not due to the toxicity of polymer or nonspecific depletion of nutrients. The ability to disrupt the bacterial communication using easy to synthesize and chemically inert polymers could provide a new concept for the development of pharmaceuticals and susceptible device coatings such as catheters.
Product Description
Supelco
N-(β-Ketocaproyl)-DL-homoserine lactone, analytical standard