MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Contamination of the norepinephrine prodrug droxidopa by dihydroxyphenylacetaldehyde.
Contamination of the norepinephrine prodrug droxidopa by dihydroxyphenylacetaldehyde.
Clinical chemistry (2010-03-09)
Courtney Holmes, Noel Whittaker, Jorge Heredia-Moya, David S Goldstein
ABSTRACT
L-threo-3,4-dihydroxyphenylserine (L-DOPS, droxidopa) is a norepinephrine (NE) prodrug under development to treat orthostatic hypotension. 3,4-Dihydroxyphenylacetaldehyde (DOPAL), an endogenous catecholaldehyde produced by enzymatic oxidative deamination of dopamine, is toxic to catecholaminergic neurons. Based on the observation of increasing plasma DOPAL after oral administration of L-DOPS to a patient, we examined whether other subjects also had DOPAL in their plasma after droxidopa administration, and whether droxidopa is contaminated with DOPAL. Thirteen subjects took 400 mg droxidopa orally. We sampled venous blood at baseline and 1, 2, 3, 6, 24, and 48 h after drug administration and assayed L-DOPS, NE, and DOPAL by use of liquid chromatography with electrochemical detection (LC-ED). Droxidopa in acidic solution (20:80 mixture of 0.04 mol/L phosphoric acid:0.20 mol/L acetic acid) was vacuum centrifuged for 1 h at 30 degrees C and then assayed by LC-ED. Droxidopa contained 0.01% DOPAL. At 6 h after droxidopa, all subjects had detectable DOPAL in plasma (1.89 nmol/L, P = 0.0001). Across the sampling times, plasma DOPAL correlated with plasma L-DOPS (r = 0.996). The mean increment in plasma DOPAL was more than 4 times that in plasma NE (0.39 nmol/L). In 2 patients with Parkinson disease and orthostatic hypotension, DOPAL was detected in plasma at baseline (0.12 nmol/L) and increased by about 70-fold after droxidopa. Vacuum concentration of droxidopa in the acid solution converted L-DOPS to DOPAL completely. Droxidopa is contaminated with DOPAL. After oral droxidopa administration, DOPAL is detected in plasma of humans. Droxidopa is susceptible to extensive nonenzymatic conversion to DOPAL.
Sigma-Aldrich
L-DOPS, ≥98% (HPLC)