MilliporeSigma
  • Structure-activity relationship of indolicidin, a Trp-rich antibacterial peptide.
Structure-activity relationship of indolicidin, a Trp-rich antibacterial peptide.
Journal of peptide science : an official publication of the European Peptide Society (2010-03-03)
Setsuko Ando, Keitarou Mitsuyasu, Yoshitake Soeda, Mariko Hidaka, Yuki Ito, Kouki Matsubara, Mitsuno Shindo, Yoshiki Uchida, Haruhiko Aoyagi
ABSTRACT
A series of Trp and Arg analogs of antibacterial indolicidin (Ind) was synthesized and the antimicrobial and hemolytic activities were investigated. [L(9)]Ind, [L(11)]Ind, [K(8),L(9)]Ind and [K(6, 8),L(9)]Ind showed desirable characteristics, exhibiting negligible hemolytic activity while keeping strong antibacterial activity. The results indicated that the Trp residue at position 11 essentially contributes to both activities and one can not be exchanged for the other, whereas the Trp residues at positions 4 and 9 play important roles in antimicrobial and hemolytic activities, respectively. The Trp residues at positions 6 and 8 play no important roles in biological activities. We then found that the retro analog of Ind showed higher antibacterial activity than Ind against both Gram-positive and Gram-negative bacteria but remarkably lower hemolytic activity than that of Ind.