MilliporeSigma
  • Toxicity and bioconcentration evaluation of RDX and HMX using sheepshead minnows in water exposures.
Toxicity and bioconcentration evaluation of RDX and HMX using sheepshead minnows in water exposures.
Ecotoxicology and environmental safety (2010-03-02)
Guilherme R Lotufo, Alfreda B Gibson, J Leslie Yoo
ABSTRACT
Lethal effects of the explosives RDX and HMX were assessed using ten-day water exposures to juvenile sheepshead minnows (Cyprinodon variegatus). For RDX, maximum mortality occurred during the first two days of exposure with a 10-d median lethal concentration (LC50) of 9.9 mg L(-1). The RDX 10-d median lethal residue (LR50) was 9.6 mg kg(-1) (34.9 μmol kg(-1)) wet weight (ww), the first RDX critical body residue reported for fish. Previous investigations reported that RDX body residues in marine amphipods up to 96 μmol kg(-1) ww and in marine mussels up to 86 μmol kg(-1) ww failed to result in significant mortality. The highest HMX concentration tested, corresponding to its apparent solubility limit in seawater (2.0 mg L(-1)), and the associated mean body residue (3 mg kg(-1) or 14 μmol kg(-1) ww) resulted in no significant mortality for exposed minnows. The mean 10-d bioconcentration factors for RDX (0.6-0.9 L kg(-1)) and HMX (0.3-1.6 L kg(-1)) were typically lower than 1, reflecting the low bioaccumulative potential for these compounds.