MilliporeSigma
[The carcinogenic and anticarcinogenic properties of neozon D].
Voprosy onkologii (1991-01-01)
G B Pliss, N N Vlasov, M A Zabezhinskiĭ
ABSTRACT
Continuous administration of neozone D to dogs and rats failed to reveal its carcinogenicity. Nor did the agent potentiate the carcinogenic effect of 2-naphthylamine and benzidine in rats. Conversely, neozone D was found to inhibit benzidine-induced carcinogenesis in female rats.
Sigma-Aldrich
N-Phenyl-2-naphthylamine, 97%