MilliporeSigma
  • Synthesis, characterization and cytotoxic activities of the [RuCl2(NO)(dppp)(L)]PF6 complexes.
Synthesis, characterization and cytotoxic activities of the [RuCl2(NO)(dppp)(L)]PF6 complexes.
Journal of inorganic biochemistry (2010-02-02)
Camilla C Golfeto, Gustavo Von Poelhsitz, Heloísa S Selistre-de-Araújo, Márcio P de Araujo, Javier Ellena, Eduardo E Castellano, Luiz G L Lopes, Icaro S Moreira, Alzir A Batista
ABSTRACT
The synthesis and characterization of ruthenium compounds of the type [RuCl(2)(NO)(dppp)(L)]PF(6) [dppp=1,3-bis(diphenylphosphino)propane; L=pyridine, 4-methylpyridine, 4-phenylpyridine and dimethyl sulfoxide] are described. The complexes were characterized by elemental analysis, UV/Vis and infrared spectroscopy, cyclic voltammetry, and X-ray crystallography for the complexes with the pyridine and 4-methylpyridine ligands. In vitro evaluation of these nitrosyl complexes revealed cytotoxic activity from 7.1 to 19.0 microM against the MDA-MB-231 breast tumor cells and showed that, in this case, they are more active than the reference metallodrug cisplatin. The 1,3-bis(diphenylphosphino)propane and the N-heterocyclic ligands alone failed to show cytotoxic activities at the concentrations tested (maximum concentration utilized=200 microM).
Sigma-Aldrich
4-Phenylpyridine, 97%