MilliporeSigma
  • Premature onset of mitosis and potentiation of chromosome damage induced by poly-D-lysine in plant cells: evidence for G2 repair.
Premature onset of mitosis and potentiation of chromosome damage induced by poly-D-lysine in plant cells: evidence for G2 repair.
Mutation research (1991-03-01)
F Cortés, S Mateos
ABSTRACT
Poly-D-lysine has been reported to induce a triggering of mitosis in plant cells due to a selective stimulatory effect on cells arrested in G2. Root-tip cells of Allium cepa L. were first exposed to maleic hydrazide (MH) early in the cell cycle and posttreated with different concentrations of the polycationic agent while in G2. The result was a dose-dependent potentiation of chromosome damage observed at metaphase without any apparent effect induced by poly-D-lysine itself. The enhancement of the yield of chromosomal aberrations was concomitant with an increase in the frequency of mitosis. In order to test further the stimulatory effect of poly-D-lysine on mitosis, as well as the consequences of a shortening of the time available for repair, cells synchronized by protracted treatment with 5-aminouracil (5-AU), which also induces chromosome damage, were allowed to recover in the presence of the polycationic compound. Our data show that a premature arrival at mitosis resulted in an increase in the frequency of damaged cells observed.
Sigma-Aldrich
5-Aminouracil, 98%