MilliporeSigma
Methsuximide for intractable childhood seizures.
Pediatrics (1991-02-01)
M B Tennison, R S Greenwood, M V Miles
ABSTRACT
Methsuximide was added to the therapeutic regimens of 25 children with intractable epilepsy. In 15 patients the drug was well tolerated and resulted in a 50% or greater reduction in seizure frequency. No serious or irreversible adverse effects were seen. Methsuximide is frequently overlooked and may be an effective adjunctive antiepileptic for children with intractable seizures.