MilliporeSigma
  • Photometric or fluorometric assay of cathepsin B, L and H and papain using substrates with an aminotrifluoromethylcoumarin leaving group.
Photometric or fluorometric assay of cathepsin B, L and H and papain using substrates with an aminotrifluoromethylcoumarin leaving group.
Biochimica et biophysica acta (1991-01-08)
J R Tchoupe, T Moreau, F Gauthier, J G Bieth
ABSTRACT
N-trifluoromethylcoumarinylamide derivatives of benzyloxycarbonyl-Arg-Arg, benzyloxycarbonyl-Phe-Arg and Arg are convenient chromogenic and fluorogenic substrates of cathepsin B, L and H, respectively. Benzyloxycarbonyl-Phe-Arg-N-trifluoromethylcoumarinylamide is also a highly sensitive substrate for papain.
Sigma-Aldrich
7-Amino-4-(trifluoromethyl)coumarin, ≥99%