MilliporeSigma
  • Antiasthmatic activity of luteolin-7-O-glucoside from Ailanthus altissima through the downregulation of T helper 2 cytokine expression and inhibition of prostaglandin E2 production in an ovalbumin-induced asthma model.
Antiasthmatic activity of luteolin-7-O-glucoside from Ailanthus altissima through the downregulation of T helper 2 cytokine expression and inhibition of prostaglandin E2 production in an ovalbumin-induced asthma model.
Biological & pharmaceutical bulletin (2009-09-02)
Meihua Jin, Ju Hye Yang, Eunkyung Lee, Yue Lu, Soonyoul Kwon, Kun Ho Son, Jong Keun Son, Hyeun Wook Chang
ABSTRACT
Previously, we reported that an ethanol extract of Ailanthus altissima has antiinflammatory activity in an ovalbumin (OVA)-sensitized murine asthmatic model. To determine the biological compounds from this plant, luteolin-7-O-glucoside (L7G) was isolated and its antiasthmatic activity was evaluated in an in vivo murine asthmatic model. L7G (10 to 100 mg/kg, per os (p.o.)) reduced the amount of eosinophil infiltration in bronchoalveolar lavage (BAL) fluid in a dose-dependent manner. In comparison, dexamethasone (5 mg/kg, p.o.), which was used as a positive control, also strongly inhibited the number of infiltrating eosinophils. L7G inhibited both the prostaglandin E(2) (PGE(2)) and serum immunoglobulin E level in BAL fluid in a dose-dependent manner. In addition, L7G inhibited the transcript profiles of interleukin (IL)-4, IL-5, and IL-13 mRNA expression levels in the murine asthma model, as determined using reverse transcription-polymerase chain reaction (RT-PCR). These results suggest that the antiasthmatic activity of L7G in OVA-induced lung inflammation may occur in part via the downregulation of T helper 2 cytokine transcripts as well as the inhibition of PGE(2) production.
Product Description
Supelco
Luteolin 7-glucoside, analytical standard
Luteolin 7-glucoside, primary reference standard
Sigma-Aldrich
Luteolin 7-O-β-D-glucoside, ≥98.0% (HPLC)