MilliporeSigma
  • The crystal structure of the Escherichia coli autoinducer-2 processing protein LsrF.
The crystal structure of the Escherichia coli autoinducer-2 processing protein LsrF.
PloS one (2009-08-29)
Zamia Diaz, Karina B Xavier, Stephen T Miller
ABSTRACT
Many bacteria produce and respond to the quorum sensing signal autoinducer-2 (AI-2). Escherichia coli and Salmonella typhimurium are among the species with the lsr operon, an operon containing AI-2 transport and processing genes that are up regulated in response to AI-2. One of the Lsr proteins, LsrF, has been implicated in processing the phosphorylated form of AI-2. Here, we present the structure of LsrF, unliganded and in complex with two phospho-AI-2 analogues, ribose-5-phosphate and ribulose-5-phosphate. The crystal structure shows that LsrF is a decamer of (alphabeta)(8)-barrels that exhibit a previously unseen N-terminal domain swap and have high structural homology with aldolases that process phosphorylated sugars. Ligand binding sites and key catalytic residues are structurally conserved, strongly implicating LsrF as a class I aldolase.
Product Description
Supelco
N-(β-Ketocaproyl)-DL-homoserine lactone, analytical standard