MilliporeSigma
  • The protozoan parasite Theileria annulata alters the differentiation state of the infected macrophage and suppresses musculoaponeurotic fibrosarcoma oncogene (MAF) transcription factors.
The protozoan parasite Theileria annulata alters the differentiation state of the infected macrophage and suppresses musculoaponeurotic fibrosarcoma oncogene (MAF) transcription factors.
International journal for parasitology (2009-03-24)
Kirsty Jensen, Giles D Makins, Anna Kaliszewska, Martin J Hulme, Edith Paxton, Elizabeth J Glass
ABSTRACT
The tick-borne protozoan parasite Theileria annulata causes a debilitating disease of cattle called Tropical Theileriosis. The parasite predominantly invades bovine macrophages (m phi) and induces host cell transformation by a mechanism that has not been fully elucidated. Infection is associated with loss of characteristic m phi functions and phenotypic markers, indicative of host cell de-differentiation. We have investigated the effect of T. annulata infection on the expression of the m phi differentiation marker c-maf. The up-regulation of c-maf mRNA levels observed during bovine monocyte differentiation to m phi was suppressed by T. annulata infection. Furthermore, mRNA levels for c-maf and the closely related transcription factor mafB were significantly lower in established T. annulata-infected cell-lines than in bovine monocyte-derived m phi. Treatment of T. annulata-infected cells with the theileriacidal drug buparvaquone induced up-regulation of c-maf and mafB, which correlated with altered expression of down-stream target genes, e.g. up-regulation of integrin B7 and down-regulation of IL12A. Furthermore, T. annulata infection is associated with the suppression of the transcription factors, Pu.1 and RUNX1, and colony stimulating factor 1 receptor (CSF1R) which are also involved in the regulation of monocyte/m phi differentiation. We believe these results provide the first direct evidence that T. annulata modulates the host m phi differentiation state, which may diminish the defence capabilities of the infected cell and/or promote cell proliferation. Musculoaponeurotic fibrosarcoma oncogene (MAF) transcription factors play an important role in cell proliferation, differentiation and survival; therefore, regulation of these genes may be a major mechanism employed by T. annulata to survive within the infected m phi.