MilliporeSigma
  • Phase II study of 3-AP Triapine in patients with recurrent or metastatic head and neck squamous cell carcinoma.
Phase II study of 3-AP Triapine in patients with recurrent or metastatic head and neck squamous cell carcinoma.
Annals of oncology : official journal of the European Society for Medical Oncology (2009-02-28)
C M Nutting, C M L van Herpen, A B Miah, S A Bhide, J-P Machiels, J Buter, C Kelly, D de Raucourt, K J Harrington
ABSTRACT
Treatment options for recurrent or metastatic head and neck squamous cell carcinoma (HNSCC) are limited with response rates to cytotoxic chemotherapy of approximately 30% and median survival of 6 months. In a multicentre phase II study, 32 patients with recurrent or metastatic HNSCC received 3-AP Triapine (3-aminopyridine-2-carboxaldehyde thiosemicarbazone), an inhibitor of ribonucleotide reductase, 96 mg/m2, daily for 4 days every 14 days (one cycle). Eligibility criteria required Eastern Cooperative Oncology Group performance status (ECOG PS) of zero to two with a life expectancy of >3 months; one prior chemotherapy regimen was allowed. Thirty patients were assessable for response and toxicity. Median age was 57 years (range 36-79) and median ECOG PS was one (range 0-2). Thirteen patients had previously been treated with chemotherapy. A total of 130 cycles were administered with a median number of cycles of 3.5 (range 1-8). Mild anaemia (40%), nausea (22%) and fatigue (22%) were commonly reported with G3 and G4 neutropenia documented in 22% and 22%, respectively. Overall response rate was 5.9% (95% confidence interval 0.2% to 28.7%). One patient achieved a partial response, eight had stable disease and 21 progressive disease. Median time to disease progression was 3.9 months. 3-AP Triapine as a single agent, at this dose and schedule, is well tolerated but has only minor activity in the treatment of advanced HNSCC.
Sigma-Aldrich
3-AP, ≥98% (HPLC)