MilliporeSigma
Zotepine: a clinical review.
Expert opinion on drug metabolism & toxicology (2009-02-10)
Ben Green
ABSTRACT
Zotepine is a second generation antipsychotic drug, a substituted dibenzothiepine tricyclic molecule, with effects on dopamine, serotonin and noradrenaline receptors. This article reviews the relevant research studies on the drug based on a search of numerous medical databases for research and literature concerning zotepine and adverse reactions reported to the UK Medicines and Healthcare Products Regulatory Agency. There are double-blind trials demonstrating zotepine as an efficacious antipsychotic with a safe side effect profile that compares favourably with other atypical antipsychotics with regard to metabolic effects and movement disorders. There are some open label studies suggesting usefulness in treatment resistant schizophrenia, but larger, double-blind studies are required.
Sigma-Aldrich
Zotepine