MilliporeSigma
  • Dietary variables associated with DNA N7-methylguanine levels and O6-alkylguanine DNA-alkyltransferase activity in human colorectal mucosa.
Dietary variables associated with DNA N7-methylguanine levels and O6-alkylguanine DNA-alkyltransferase activity in human colorectal mucosa.
Carcinogenesis (2009-01-27)
H A Billson, K L Harrison, N P Lees, C N Hall, G P Margison, A C Povey
ABSTRACT
Components of human diets may influence the incidence of colorectal adenomas, by modifying exposure or susceptibility to DNA-damaging alkylating agents. To examine this hypothesis, a food frequency questionnaire was used to assess the diet of patients recruited for a case-referent study where biopsies of normal colorectal mucosa were collected during colonoscopy and subsequently analysed for DNA N7-methylguanine (N7-MeG) levels, as an indicator of exposure, and activity of the DNA repair protein O6-alkylguanine DNA-alkyltransferase (MGMT), as an indicator of potential susceptibility. Cases with histologically proven colorectal adenomas (n = 38) were compared with referents (n = 35) free of gastrointestinal neoplasia. The case group consumed significantly more red meat (4.5 versus 3.4 servings/week, P < 0.05), processed meats, (4.7 versus 3.2 servings/week, P < 0.05) and % food energy as fat (34.9 versus 30.7%, P < 0.001). N7-MeG [mean: 95% confidence interval (CI)] levels were significantly lower in the group that consumed the highest proportion of dietary fibre/1000 kcal in comparison with the group with the lowest intake (0.61; 0.35-0.86 versus 1.88; 0.88-2.64 micromol/mol dG, P < 0.05). N7-MeG levels were also inversely associated with folate consumption (P < 0.05). MGMT activity (mean; 95% CI) was significantly higher in the group with the lowest consumption of vegetables than in the group with the greatest vegetable consumption (7.02; 5.70-8.33 versus 4.93; 3.95-5.91 fmol/microg DNA, P < 0.05). Our results are consistent with the hypothesis that dietary factors may modify exposure or susceptibility, respectively, to DNA damage by alkylating agents.
Sigma-Aldrich
7-Methylguanine, ≥98.0% (HPLC)