MilliporeSigma
Advanced substrate fabrication for cell microarrays.
Biomacromolecules (2009-01-23)
Andrew L Hook, Helmut Thissen, Nicolas H Voelcker
ABSTRACT
The fabrication and characterization of chemical patterns using a technique that can be readily integrated with methods currently used for the formation of microarrays is presented. A high density poly(ethylene glycol) coating was deposited on glass slides as a background exhibiting low cell attachment properties. Phenylazide modified polymers were then printed on this background. UV irradiation of these polymer arrays resulted in the cross-linking of the polymer spots and their covalent attachment to the surface. Cell attachment was shown to follow the resultant surface chemistry pattern. Furthermore, the use of a robotic contact printer enabled the facile deposition of DNA microarrays on top of and aligned with the polymer microarrays. A transfected cell microarray was generated in this way, demonstrating not only the ability of this platform to limit cell attachment to specific regions, but the suitability for chip-based functional genomics, in particular, and high density cell assays in general.
Sigma-Aldrich
Azidobenzene solution, ~0.5 M in tert-butyl methyl ether
Sigma-Aldrich
Azidobenzene solution, ~0.5 M in 2-methyltetrahydrofuran, ≥95.0% (HPLC)