MilliporeSigma
  • [Conversion of 17 alpha-methyltestosterone to methandrostenolone by the bacterium Pimelobacter simplex VKPM Ac-1632 with the presence of cyclodextrins].
[Conversion of 17 alpha-methyltestosterone to methandrostenolone by the bacterium Pimelobacter simplex VKPM Ac-1632 with the presence of cyclodextrins].
Prikladnaia biokhimiia i mikrobiologiia (2009-01-17)
A V Druzhinina, V A Andriushina, T S Stytsenko, N E Voĭshvillo
ABSTRACT
Conditions of conversion of 17 alpha-methyltestosterone to methandrostenolone with the presence of modified beta-cyclodextrins (methylcyclodextrin, hydroxypropylcyclodextrin, and hydroxyethylcyclodextrin) in the steroid:cyclodextrin ratio 1:1 were studied. The experimental solutions of modified beta-cyclodextrins were prepared in deionized water with 5-7% methanol. Under the conditions found to be optimal, 1,2-dehydrogenation of 17 alpha-methyltestosterone was carried out with 2-4 g/l Pimelobacter simplex VKPM Ac-1632 biomass. At the substrate concentration 5-20 g/l, the reaction occurred for 1-15 h without any by-products. The maximum rate of methandrostenolone accumulation was observed with hydroxypropylcyclodextrin. The methylcyclodextrin solution can be reused for complete 17 alpha-methyltestosterone conversion at the concentration 5 g/l.
Product Description
Supelco
Methandienone solution, 1.0 mg/mL in 1,2-dimethoxyethane, ampule of 1 mL, certified reference material, Cerilliant®