MilliporeSigma
  • Myocardial resistance to ischemic and reperfusion injuries under conditions of chronic administration of opioid receptor agonists and antagonists.
Myocardial resistance to ischemic and reperfusion injuries under conditions of chronic administration of opioid receptor agonists and antagonists.
Bulletin of experimental biology and medicine (2008-12-27)
Yu B Lishmanov, D L Stakheev, A V Krylatov, N V Naryzhnaya, L N Maslov, M V Ovchinnikov, F Kolar
ABSTRACT
Chronic treatment with opioid receptor ligands: nonselective peptide opioid receptor agonist dalargin (intraperitoneally in a dose of 1 mg/kg), selective nonpeptide kappa-receptor agonist GR 89696 (subcutaneously in a dose of 0.03 mg/kg), nonselectrive nonpeptide antagonist quadazocine (subcutaneously in a dose of 3 mg/kg) or naltrexone (subcutaneously in a dose of 10 mg/kg) for 20 day had no effect of the incidence of ischemic ventricular arrhythmias and the size of necrotic zone after coronary occlusion and reperfusion in rats in vivo.
Sigma-Aldrich
GR 89696 fumarate salt, solid