MilliporeSigma
  • Enantioselective sequential conjugate addition-allylation reactions: a concise total synthesis of (+)-podophyllotoxin.
Enantioselective sequential conjugate addition-allylation reactions: a concise total synthesis of (+)-podophyllotoxin.
Organic letters (2008-12-25)
Yingming Wu, Jingfeng Zhao, Jingbo Chen, Chengxue Pan, Liang Li, Hongbin Zhang
ABSTRACT
A highly flexible and concise total synthesis of (+)-podophyllotoxin featured with an enantioselective sequential conjugate addition-allylation reaction was reported. Starting from commercially available 3,4,5-trimethoxycinnamic acid, this new route leads to (+)-podophyllotoxin 1 in only eight steps with 29% overall yield.