MilliporeSigma
  • Biological measure of compliance to Artesunate plus Amodiaquine association: interest in a Mono-Desethyl-Amodiaquine blood assay?
Biological measure of compliance to Artesunate plus Amodiaquine association: interest in a Mono-Desethyl-Amodiaquine blood assay?
Acta tropica (2008-12-06)
Sophie Sarrassat, Madiagne Sakho, Jean Yves Le Hesran
ABSTRACT
The deployment of Artemisinin-based Combination Therapy for treating uncomplicated malaria poses problems in the patient compliance to these new treatments. The aim of our study was to investigate the relationship between compliance to 3 days treatment with Artesunate plus Amodiaquine (AS+AQ) and the Mono-Desethyl-Amodiaquine (MDA) blood concentration on the fourth day. A reference scale of mean MDA blood concentrations was constructed in 40 healthy adults. Each concentration corresponded to the MDA level on day 3 in a subject having one of the seven compliance degrees defined by the number and sequence of drug intakes from day 0 to day 2: one single dose on day 0, day 1 or day 2; two single doses separated by 24h, on day 0 and day 1 or on day 1 and day 2; two single doses separated by 48 h, on day 0 and day 2; three single doses, on day 0, day 1 and day 2. MDA was assayed in whole blood samples by HPLC. Non-parametric Mann and Whitney U tests were used for the comparison of two means. Our results demonstrated no clear relationship between the mean MDA blood concentrations on day 3 and compliance degrees, according to neither the number nor the sequence of doses taken. In particular, even though the differences were not significant, the mean concentration after three doses, expected to be the maximum, was unexpectedly lower than after two doses, on day 0 and day 1 or on day 1 and day 2. The high inter-individual variability of MDA concentrations attributed to the different rates of hepatic metabolism of each individual appears to have a greater effect on MDA levels than the number or timing of doses. Therefore, it seems that the role of a MDA blood assay is limited in use to discerning if none or one or more doses have been taken. A MDA assay do not allow to measure the compliance degree of one patient to AS+AQ association. Presently, interview and pill count following treatment seem to be the only tools available that may permit differentiation between degrees of compliance.
Product Description
Supelco
N-Desethylamodiaquine dihydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®