MilliporeSigma
  • Genepolide, a sesterpene gamma-lactone with a novel carbon skeleton from mountain wormwood (Artemisia umbelliformis).
Genepolide, a sesterpene gamma-lactone with a novel carbon skeleton from mountain wormwood (Artemisia umbelliformis).
Journal of natural products (2008-12-05)
Giovanni Appendino, Orazio Taglialatela-Scafati, Adriana Romano, Federica Pollastro, Cristina Avonto, Patrizia Rubiolo
ABSTRACT
The sesterpene gamma-lactone genepolide (5) has been isolated from a Swiss horticultural variety of mountain wormwood (Artemisia umbelliformis) developed as a thujones-free alternative to native Western Alps wormwoods for the production of liqueurs. Genepolide is the formal Diels-Alder adduct of the exomethylene-gamma-lactone costunolide (2) and the diene myrcene (6), two poorly reactive partners in cycloaddition reactions, and its structure was elucidated through a combination of spectroscopic methods. An investigation on the thermal stability of mixtures of 2 and 6, as well as considerations on the sensitivity of 2 to Brønsted and Lewis acids, suggests that 5 is a genuine natural product and that the Swiss chemotype of A. umbelliformis contains Diels-Alderase enzymatic activity that is lacking in native mountain wormwoods from Western Alps. Remarkable differences in thermal and acid-catalyzed reactions of the cyclodecadiene moiety of 2 and 5 suggest that quaternarization at C-11 has far-reaching effects on the reactivity of their homoconjugated medium-sized diene system. The wide occurrence of this structural motif in sesquiterpenoids makes this issue worth a systematic investigation.
Sigma-Aldrich
Costunolide, ≥97% (HPLC)