MilliporeSigma
  • Highly efficient copper-catalyzed cascade synthesis of quinazoline and quinazolinone derivatives.
Highly efficient copper-catalyzed cascade synthesis of quinazoline and quinazolinone derivatives.
Chemical communications (Cambridge, England) (2008-12-03)
Cheng Huang, Yuan Fu, Hua Fu, Yuyang Jiang, Yufen Zhao
ABSTRACT
We have developed a general and highly efficient copper-catalyzed method for synthesis of quinazoline and quinazolinone derivatives, the target products were obtained in good to excellent yields via cascade reactions of amidine hydrochlorides with substituted 2-halobenzaldehydes, 2-halophenylketones, or methyl 2-halobenzoates, and the method is of simple, economical and practical advantages.
Sigma-Aldrich
Acetamidine hydrochloride, 95%